Melting furnaces, K 1/10 - K 4/13 and KC 1/15 - KC 2/15
Supplier: Nabertherm
NABE001420780EA 17260 EUR
NABE001420780 NABE001420127 NABE001420730 NABE001420427 NABE001420527 NABE001420227 NABE001420327 NABE001420627
Melting furnaces, K 1/10 - K 4/13 and KC 1/15 - KC 2/15
These compact melting furnaces for melting of non-ferrous metals and alloys are one of a kind and have a number of technical advantages. Designed as tabletop models, they can be used for many laboratory applications. The practical counter balanced hinge with shock absorbers and the spout (not KC) on the front of the furnace make exact dosing easy when pouring the melt.
  • Tmax 1000 °C, 1300 °C or 1500 °C with melt temperatures about 80 °C to 110 °C lower
  • Crucible sizes of 0,75, 1,5 and 3 litres
  • Crucible with integrated pouring spout of iso-graphite included with delivery
  • Additional spout (not KC) mounted at the furnace for exact pouring
  • Compact bench-top design, simple emptying of crucible by tilting system with gas damper
  • Crucible for heating up of melting furnace insulated with a hinged lid, lid opened when pouring
The K 1/10 - K 4/13 is equipped with the simple R7 controller whereas the KC furnaces are equipped with a 3508 controller. Other crucible types available on request, e.g. steel. Additional equipment: observation hole for melt, over-temperature limiter for the furnace chamber with automatic reset to protect against overtempearture. The limit controller switches off the heating when the pre-set limit temperature has been reached and does not switch it on again until the temperature fallsbelow the setting again.