Organic reference standard, Deoxynivalenol-d1 solution, analytical standard, Supelco®
Supplier: Merck
34155-2MLEA 539 EUR
34155-2ML
Organic reference standard, Deoxynivalenol-d1 solution, analytical standard, Supelco®
Standards Organic Standards
Deoxynivalenol-d1 solution can be used as working standard in the analysis of trichothecenes and zearalenones in grain cereal, flour and bread using liquid chromatography coupled with mass spectrometry (LC/MS-MS). It can also be used as an internal standard in the determination of mycotoxins in baby foods using liquid chromatography coupled with mass spectrometry (LC/MS-MS).