Hydrocarbon test mix, in chloroform (varied conc.), certified reference material, Supelco®
Supplier: Merck
TraceCERT®
48244.EA 43.8 EUR
48244.
Hydrocarbon test mix, in chloroform (varied conc.), certified reference material, Supelco®
Standards Petroleum Industry Standards
An ideal mix for checking column installation when you use a capillary column. This product is also used to determine theoretical plates.
Analytes:
Dodecane; Tridecane; Tetradecane; Pentadecane; Hexadecane; Heptadecane
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.