The arch and slot style hose racks accommodate a variety of hoses, and are available in standard or mini sizes. These wall-mounted racks keep various size hoses organized and off the floor.
- Extends hose working life and ensures 100% drainage
- Compact designs ideal for use in any cleanroom environment
- Eliminates clutter and enhances safety
- Conveniently stores sanitary gaskets, clamps, and adaptors in one location (not included on Arch Rack Mini)
- Constructed from 304 electropolished stainless steel
- Ideal for biopharmaceutical cleanroom applications
Three unique wall-mounted hose rack styles are available to choose from, including slot, arch, and mini-arch shaped designs.
Certifications: Meets FDA and 3A regulations for hose storage, arch and slot style racks suits cGMP.
Delivery information: Supplied with mounting hardware.