Hexane (mixture of isomers) ≥85%, CHROMASOLV™ for HPLC

Supplier: Honeywell Chemicals
Danger

Synonyms: HexanesIsohexane

34994-2.5LEA 240.07 EUR
34994-2.5L
Hexane (mixture of isomers) ≥85%, CHROMASOLV™ for HPLC
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
MW: 86,18 g/mol
Boiling Pt: 68,7 °C
Melting Pt: –95 °C
Density: 0,66 g/cm3 (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
UN: 1208
ADR: 3,II
https://ie.vwr.com/stibo/web/std.lang.all/05/68/16640568.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR