Ethyl alcohol (taxed) Pure, meets analytical specification of USP (200 Proof), Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Ethyl alcohol

493546-1LEA 203.15 EUR
493546-1L 493546-2.5L
Ethyl alcohol (taxed) Pure, meets analytical specification of USP (200 Proof), Riedel-de Haën™
Ethanol

Taxed, for domestic sale in Ireland and United Kingdom only

Formula: C₂H₅OH
MW: 46,07 g/mol
Boiling Pt: 78 - 79 °C
Melting Pt: –115 °C
Density: 0,790 - 0.791 g/cm3
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Identity A complying
Identity B complying
Assay (GC) min. 99.50 Vol.%
Non-volatile matter max. 25 ppm
Acidity or alkalinity complying
Acetaldehyde & Acetale (GC) max. 10 Vol.ppm
Benzene (GC) max. 2 Vol.ppm
Methanol (GC) max. 200 Vol.ppm
Volatile impurities (GC) complying
Absorbance 270-340 nm max. 0.10
Absorbance 250-260 nm max. 0.30
Absorbance at 240 nm max. 0.40
Curve course (UV) complying
Colour of the solution complying
Clearness of the solution complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR