Dichloromethane ≥99.9% (without stabilizer, GC) stabilised, Laboratory Reagent
Supplier: Honeywell Chemicals
|
Warning
|
Synonyms:
Methylene chloride
Stabilisation:
Stabilised with ethanol 0,1-0,4%
HONC14609-5LEA
135.98
EUR
HONC14609-5L
14609-4X5L
Dichloromethane ≥99.9% (without stabilizer, GC) stabilised, Laboratory Reagent
Dichloromethane
- ≥95% for general laboratory use.
Formula:
CH₂Cl₂ MW: 84,93 g/mol Boiling Pt: 39 - 41 °C Melting Pt: -97 °C Density: 1,32 - 1.33 g/cm3 Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III |
![]() |
Specification Test Results
Assay GC (Product + Ethanol) | min. 99.9 Gew.% |
Free acid (as HCl) | max. 0.0005 % |
Water (Karl Fischer) | max. 0.01 % |
Non-volatile matter | max. 0.002 % |
Ethanol (GC) | 0.1 - 0.4 Gew.% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...