Dimethyl sulphoxide, anhydrous ≥99.9% for applications requiring low water content <0.0050% H₂O, Riedel-de Haën™
Supplier: Honeywell Chemicals
Synonyms:
DMSO
, Methyl sulphoxide
Packed in septum-sealed bottle.
HONC66206-2.5LEA
393.14
EUR
HONC66206-2.5L
HONC66206-1L
HONC66206-100ML
Dimethyl sulphoxide, anhydrous ≥99.9% for applications requiring low water content <0.0050% H₂O, Riedel-de Haën™
Dimethyl sulphoxide
Formula:
(CH₃)₂SO MW: 78,14 g/mol Boiling Pt: 189 °C Density: 1,104 g/cm³ Flash Pt: 87 °C |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...