Mop wipes, Light and Super
Supplier: Chicopee
115-2491EA 9.4 EUR
115-2491
Mop wipes, Light and Super
Wipes Disposable Wipes and Towels Wipes for General Applications
These microfibre wipes are specially designed for use with the Chicopee Fluid Mopping System.
  • Reduce cleaning time
  • Microfibre technology to help capture dirt and bacteria in the mop
  • Help to reduce the risk of cross contamination by discarding after use rather than laundering
Simply place the mopping wipe on the floor and it will attach to the mopping frame using the micro fastener. Once the wipe has become filled with dirt simply remove and throw away.
Light mop wipes are able to clean between 50 to 100 m² using the liquid on board the Chicopee Fluid Mopping System, thus allowing users to clean much larger areas than with traditional bucket impregnated mop systems.
Thanks to its unique disposable nature the Chicopee Super mop wipe can also be used to clean up spillages, its unique absorbent core can be attached to the floor mop to soak up high amounts of liquid, preventing bending down and speeding up cleaning times. Designed to clean 20 to 25 m².