You Searched For: Swabs for Cleaning

Swabs for cleaning prepare objects prior to entering cleanrooms and maintain critical environment conditions. With a soft foam head, delicate items will not be damaged during dirt and dust removal. The anti-static tip guarantees that interior charges remain neutral for ultra-sensitive testing. Compatible with common solvents, the highly absorbent swabs pick up even the most difficult contaminants. The disposable swabs for cleaning have different-shaped tips to best fit surfaces.

Swabs for cleaning prepare objects prior to entering cleanrooms and maintain critical environment conditions. With a soft foam head, delicate items will not be damaged during dirt and dust removal. The anti-static tip guarantees that interior charges remain neutral for ultra-sensitive testing. Compatible with common solvents, the highly absorbent swabs pick up even the most difficult contaminants. The disposable swabs for cleaning have different-shaped tips to best fit surfaces.


72  results were found

SearchResultCount:"72"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (710-0544)
Supplier: Texwipe
Description: Swab with open-cell PU foam head. This surrounds a cotton bud core, which provides cushioned support and additional sorbency.
UOM: 1 * 2.500 items


Supplier: Texwipe
Description: Low TOC swabs are made from 100% polyester knit materials, which are further cleaned for use in Total Organic Carbon (TOC) analysis.

Supplier: Texwipe
Description: The sterile swabs are constructed from the highest quality 100 ppi polyurethane foam. Complete thermal bond construction eliminates adhesive contamina...
Catalog Number: (710-0575)
Supplier: Texwipe
Description: Laundered open-cell CleanFoam head is firm with a pointed tip.
UOM: 1 * 500 items


Supplier: Texwipe
Description: Swab with a thermally-moulded CleanFoam head on a flexible paddle. Made from PU foam with 100 pore per inch porosity.

Supplier: Texwipe
Description: These swabs feature 100% continuous filament polyester that is heat-moulded (thermally-bonded) onto the handle, sealing exposed edges to reduced fibre...

Supplier: Texwipe
Description: Thermally-bonded swab for applications requiring high absorbency, low particle generation, and uniform dispersing of solvents.

Supplier: Texwipe
Description: Sterile cotton swabs are constructed from 100% USP grade cotton. Bonded securely to a shaft by an aqueous-based adhesive. Made to exacting and consist...

Supplier: Texwipe
Description: Small head swabs for use with a solvent such as isopropanol made from PU foam with 100 pore per inch porosity.

Catalog Number: (115-1814)
Supplier: Texwipe
Description: Sampling swabs made from the cleanest 100% polyester knit materials.
UOM: 1 * 100 items


Catalog Number: (710-0569)
Supplier: Texwipe
Description: Swab for cleaning small, confined areas made from polyurethane foam, with 100 pore per inch porosity.
UOM: 1 * 500 items


Supplier: Texwipe
Description: Thermally-bonded swabs with 100 pore per inch porosity.

Supplier: Texwipe
Description: These swabs feature a soft head with low abrasion.

Supplier: Texwipe
Description: Swab features a compressed PU head with precision pointed tip and side ridges, on a short PP handle.

Supplier: Texwipe
Description: General-purpose swabs constructed from medical grade PU foam.

Sale

Supplier: Texwipe
Description: These sets contain components for sampling different locations in the production area, and provide an efficient means of transporting the samples from...

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 16 of 72
no targeter for Bottom