You Searched For: Swabs for Sampling

Typically having heads manufactured of polyester knit fabrics, swabs for sampling have the lowest level of released particles and highest sample recovery rates. After lightly moistening, the flexible applicators can pick up residue from any shape surface before sample is eluted for analysis. The single- or double-tipped collection devices utilize a less destructive method than bulk sampling. Pre-cleaned for immediate use, the disposable sampling swabs guarantee that tests remain uncontaminated.

Typically having heads manufactured of polyester knit fabrics, swabs for sampling have the lowest level of released particles and highest sample recovery rates. After lightly moistening, the flexible applicators can pick up residue from any shape surface before sample is eluted for analysis. The single- or double-tipped collection devices utilize a less destructive method than bulk sampling. Pre-cleaned for immediate use, the disposable sampling swabs guarantee that tests remain uncontaminated.


44  results were found

SearchResultCount:"44"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR Chemicals
Description: Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sp...

MSDS Sale

Supplier: VWR Collection
Description: Disposable 'foam over cotton' tip swab features 100% polyurethane foam with 100 ppi, and a wooden shaft.

Supplier: VWR Collection
Description: Critical swabs designed for cleaning disc drives, optics, PC board assemblies and other sensitive components.

Sale

Supplier: VWR Collection
Description: Swabs feature 100 ppi open or closed cell polyurethane foam, with 100% virgin polypropylene handle. Some have a blue, glass fibre, reinforced polyprop...

Catalog Number: (MMMPRS96010DE)
Supplier: 3M Food Safety
Description: 3Mâ„¢ Swab-Sampler with 10 ml D/E neutralising Broth is a convenient and quick collection device for environmental sampling and testing over small surfa...
UOM: 1 * 100 items


Supplier: STERILIN
Description: This innovative nylon fibre technology and unbeatable sample collection and release make the swabs PCR compatible for molecular analysis.

Supplier: COPAN
Description: 4N6FLOQSwabs® Genetics is the line of Copan patented FLOQSwabs® specifically designed and dedicated to DNA collection for human identification.

Sale

Supplier: KIMA SAS DI CHIARIN RENZO & C
Description: These sterile swabs are designed with either a plastic or an aluminium stick.

Supplier: COPAN
Description: Surface control swabs and sponges designed for the hygiene monitoring of surfaces.

Catalog Number: (MMMPRS96010BPW)
Supplier: 3M Food Safety
Description: 3Mâ„¢ Swab-Sampler with 10 ml Buffered Peptone Water Broth is a convenient and quick collection device for environmental sampling and testing over small...
UOM: 1 * 100 items


Supplier: COPAN
Description: An extensive range of ready-to-use environmental swabs. The swabls are designed for use in a broad range of environments, including food production, p...
Catalog Number: (MMMPRS96010NB)
Supplier: 3M Food Safety
Description: 3Mâ„¢ Swab-sampler with 10 ml neutralising buffer is a convenient and quick collection device for environmental sampling and testing over small surface ...
UOM: 1 * 100 items

MSDS


Catalog Number: (MMMPRS9604NB)
Supplier: 3M Food Safety
Description: 3Mâ„¢ Swab-sampler with 4 ml neutralising buffer is a convenient and quick collection device for environmental sampling and testing over small surface a...
UOM: 1 * 100 items


Catalog Number: (TEXWCM502)
Supplier: Texwipe
Description: Coventryâ„¢ microtips are designed for precision applications needing pinpoint accuracy. The tapered porous polyvinylidene fluoride tip is effective as ...
UOM: 1 * 1.200 items


Catalog Number: (MMMP6433)
Supplier: 3M Food Safety
Description: The 3Mâ„¢ Quick Swab is a ready-to-use environmental swab system consisting of a five inch, rayon-tipped swab containing Letheen neutralising buffer to ...
UOM: 1 * 250 items


Catalog Number: (DELT300210)
Supplier: DELTALAB
Description: Sterile swabs sterilised by ethylene oxide for use when samples do not need to be transported.
UOM: 1 * 1.000 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 16 of 44
no targeter for Bottom