Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (8)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Facial Tissues

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (115-0600)
Supplier:  VWR Collection
Description:   Two ply facial tissues made from 100% cellulose.
UOM:  1 * 150 items
Supplier:  KIMBERLY CLARK
Description:   Extra soft 2 ply tissues, dispensed one at a time from cube dispenser.
Catalog Number: (KIMBH6252010)
Supplier:  KIMBERLY CLARK
Description:   Very soft 2 ply facial tissues.
UOM:  1 * 3.000 items
Supplier:  KIMBERLY CLARK
Description:   These soft hand towels are made from strong, highly absorbent AIRFLEX* fabric and conveniently supplied in a stylish, splash-resistant pop-up box to k...
UOM:  1 * 1 items
Catalog Number: (KIMB8837)
Supplier:  KIMBERLY CLARK
Description:   Premium quality 2-ply tissues made from 100% highly refined recycled fibres - FSC certified. Can also be used with the elegant looking chromed dispens...
UOM:  1 * 2.100 items
Catalog Number: (115-1723)
Supplier:  SCA
Description:   Premium 2 ply facial tissues made of pure cellulose.
UOM:  1 * 3.000 items
Catalog Number: (ZVGA16515)
Supplier:  Zellstoff-Vertriebs-GmbH & Co. KG
Description:   These double-layered laboratory and facial tissues from zetBox® are highly absorbent and ultra-soft. Perfect for sensitive application areas.
UOM:  1 * 4.500 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 7  of 7