Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (4)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Cloths for Delicate Tasks

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"13"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (CONTBS20CB-12)

Supplier:  CONTEC
Description:   These polyester cloths are ideal for the surface preparation prior to painting. Also suitable for particle removal in printed circuit board manufactur...
UOM:  1 * 500 items
Supplier:  Zellstoff-Vertriebs-GmbH & Co. KG
Description:   Soft non woven cloths made of Airlaid material, ideal for sensitive applications.
Supplier:  3M
Description:   This microfibre cloth is made from a blend of fibres, the structure of which ensures highly efficient and environmentally friendly cleaning. Perfect f...
Supplier:  MICRONOVA
Description:   These durable wipes are designed for medical device, pharmaceutical and hi-tech applications. The material is a latex exterior bonded with rayon, whic...
Supplier:  MICRONOVA
Description:   Combining the versatility of a high quality wipe with the security of a glove, the basic mitt will fit comfortably over a gloved hand. Available in 10...
Supplier:  MICRONOVA
Description:   These polyester wipes are double-thick and feature an inverted hem which guards against the trapping and generation of particles through exposed seams...
Supplier:  MICRONOVA
Description:   These wipes are constructed from bonded, non woven polyvinyl alcohol (PVA). The fast-drying, absorbent properties of PVA are ideal for semiconductor a...
Catalog Number: (MICRWP68-99H)

Supplier:  MICRONOVA
Description:   This wipe is made of 100% polyester covered in polyester abrasive mesh. It presents a cleanroom alternative to household grade abrasive pads. To be us...
UOM:  1 * 10 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8  of 13
  1  2  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping