You Searched For: Shaking Block Heater/Coolers
With high stability even at the maximum speed, shaking block heaters and coolers mix samples at precise temperatures. Evenly blending substances to be homogenous, for reproducible and reliable results. Machines with LCD touch screens constantly monitor the testing conditions as well as transfer collected data to a computer for further review and storage. NIST traceable certificates are offered for select shaking block heaters and coolers. The equipment works well with interchangeable blocks to house any sample container employed.
With high stability even at the maximum speed, shaking block heaters and coolers mix samples at precise temperatures. Evenly blending substances to be homogenous, for reproducible and reliable results. Machines with LCD touch screens constantly monitor the testing conditions as well as transfer collected data to a computer for further review and storage. NIST traceable certificates are offered for select shaking block heaters and coolers. The equipment works well with interchangeable blocks to house any sample container employed.
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR Collection
Description: The cooling thermal shake touch is designed for applications that require consistent, precise results. With heating, cooling and shaking capabilities,...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.