You Searched For: Water Baths, Unstirred

Unstirred water baths provide the standard convection current heating for samples that require an increased temperature environment. Preferred heat sources when working with hazardous material that cannot be exposed to a direct flame, the containers incorporate lids to trap heat within. With a stainless steel construction, the laboratory instruments prevent corrosion and are easy to clean in between uses. The unstirred water baths have analogue or digital controls to reach the temperature stability required.

Unstirred water baths provide the standard convection current heating for samples that require an increased temperature environment. Preferred heat sources when working with hazardous material that cannot be exposed to a direct flame, the containers incorporate lids to trap heat within. With a stainless steel construction, the laboratory instruments prevent corrosion and are easy to clean in between uses. The unstirred water baths have analogue or digital controls to reach the temperature stability required.


35  results were found

SearchResultCount:"35"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (462-0244)
Supplier: VWR Collection
Description: Water baths with double-walled, stainless steel (inner) and coated steel sheet (outer) housing. Temperature is evenly distributed by a flat radiator. ...
UOM: 1 * 1 items


Supplier: VWR Collection
Description: Unstirred water baths with double walled, stainless steel (inner) and coated steel sheet (outer) housing. All baths except the 2 litre models feature ...

Supplier: VWR Collection
Description: Made from stainless steel or polycarbonate. Ideal for use with VWR® immersion thermostats. For use where the durability of stainless steel or the tran...

Sale

Supplier: GRANT INSTRUMENTS LTD.
Description: Premium baths which are constructed from corrosion resistant stainless steel for durability in demanding environments. Available in five sizes: 5, 12,...

Supplier: GRANT INSTRUMENTS LTD.
Description: High quality, digital unstirred water bath range with choice of models from 2 to 34 litres and a dual bath with 5 and 12 litres. Dual baths are ideal ...

Supplier: JULABO GmbH
Description: Unstirred water bath, CORIO™, B17, Stainless steel, Capacity: 13 - 17 L, Temperature range: ≤150 °C, Bath W×D×H: 300×320×200 mm, W×D×H: 370×400×250 mm

Supplier: JULABO GmbH
Description: Unstirred water bath, CORIO™, BT27, Capacity: 20 - 27 L, Temperature range: ≤100 °C, Bath W×D×H: 300×350×200 mm

Supplier: GRANT INSTRUMENTS LTD.
Description: Digital unstirred water bath range with choice of models from 5 to 18 litres. All units have stainless steel tanks and durable painted steel cases, of...

Supplier: GRANT INSTRUMENTS LTD.
Description: Digital unstirred water bath range with choice of models from 5 to 26 litres. All units have stainless steel tanks and durable painted steel cases, of...

Supplier: JULABO GmbH
Description: PURA stainless steel water baths are functional and reliable for everyday operations due to their high material and component quality. The easy to use...

Catalog Number: (442-0814)
Supplier: 2MAG
Description: Stainless steel mixBATH, durable, robust, is suitable for MIXdrive 6/15/60 as well as MIXdrive 1/1 eco/1 XS, fitting for immersion thermostats (heatin...
UOM: 1 * 1 items


Catalog Number: (JULA9461701)
Supplier: JULABO GmbH
Description: Open bath circulator with high quality bath tanks made of transparent plastic or stainless steel.
UOM: 1 * 1 items

New Product


Supplier: LAUDA
Description: Simple to operate water baths. Working temperature range is 25 to 100 °C.

Supplier: LAUDA
Description: Unstirred water bath, PC, Capacity: 20 L, Temperature range: ≤100 °C, Bath W×D×H: 322×506×208 mm

Supplier: LAUDA
Description: Insulated stainless steel baths which can be used up to 200 °C. The outer jacket is made of powder coated sheet steel. All baths are equipped with a b...

Supplier: IKA
Description: Unstirred water bath, IB R 9 eco, PC, Capacity: 9 L, Temperature range: ≤100 °C, Bath W×D×H: 307×239×151 mm, W×D×H: 356×287×167 mm

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 16 of 35
no targeter for Bottom