You Searched For: White Light Transilluminators
Compact white light transilluminators with brilliant backlit illumination are used for viewing Coomassie Blue- or silver-stained gels. Uniform white light diffusion enables viewing of autogradiographs, X-ray film, film positives, negatives, and microtiterplates via highly sensitive detection methods. White light transilluminators are built from durable parts and sealed into a unit for long-lasting service life. These instruments are designed for increased uniformity, instant-on capabilities, no lamp flicker, and reduced electrical consumption. Ideal for small workspaces or laboratories.
Compact white light transilluminators with brilliant backlit illumination are used for viewing Coomassie Blue- or silver-stained gels. Uniform white light diffusion enables viewing of autogradiographs, X-ray film, film positives, negatives, and microtiterplates via highly sensitive detection methods. White light transilluminators are built from durable parts and sealed into a unit for long-lasting service life. These instruments are designed for increased uniformity, instant-on capabilities, no lamp flicker, and reduced electrical consumption. Ideal for small workspaces or laboratories.
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (730-1002)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: White Light transilluminators offer uniform Light diffusion for autoradiographs, negatives, X-ray film, film positives, microtitre plates and gels.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.