You Searched For: Gel Dryers, Air Drying
Gel drying frames allow researchers to easily handle dry gels crack-free without the use of heat/vacuum dryers. Each unit is light and robust, and functions with slab, gradient, 2D, immunodiffusion, immunoelectrophoretic, and enzyme activity gels. Gels can be air-dried or through the application of heat for heavier gels. Colour of stains for proteins, glycoproteins, nucleic acids, and colorized enzymatic products remains stable for years. Drying envelopes/cellophane sheets are transparent, flat, and wrinkle-free.
Gel drying frames allow researchers to easily handle dry gels crack-free without the use of heat/vacuum dryers. Each unit is light and robust, and functions with slab, gradient, 2D, immunodiffusion, immunoelectrophoretic, and enzyme activity gels. Gels can be air-dried or through the application of heat for heavier gels. Colour of stains for proteins, glycoproteins, nucleic acids, and colorized enzymatic products remains stable for years. Drying envelopes/cellophane sheets are transparent, flat, and wrinkle-free.
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View
Rate These Search Results
Supplier: OWL SCIENTIFIC
Description: Gel drying frame, minigel (10×10 cm), Gel size (W×L): 140×140 mm
Supplier: DIVERSIFIED BIOTECH
Description: Gel drying frames allow researchers for easy handling and crack free dry gels without the use of heat/vacuum dryers.
Supplier: Serva
Description: Gel drying frame kit suitable for drying mini vertical slab gels up to 100×100 mm.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 3 of 3
no targeter for Bottom