Rotational viscometers, Smart
Supplier: Fungilab
Smart
FUNGV211002NEA 4170 EUR
FUNGV211002N FUNGV211001N 645-0083 645-0082 645-0084 FUNGV211003N
Rotational viscometers, Smart
Viscometers Rotational Viscometers
Rotational viscometers for routine measurements in quality control and research and development laboratories.
  • Precision of ±1,0% full scale and resolution of 0,1 for viscosity range lower than 10000 cP
  • Time measurement functions, time to torque and time to stop with ten working memories
  • AUTO-TEST, AUTO-RANGE functions
  • AISI 316 stainless steel spindles - marked and easily identifiable
  • USB interface allows data transfer to a PC in Excel® format using the data logger software
The six touch key user interface enables the user to easily navigate the program menu and decide on what is included in the readout of the monochrome graphic display.
The readout includes selected speed; selected spindle; viscosity reading (either cP or cSt); shear rate and shear stress (with coaxial spindles). Temperature data can also be displayed if the PT100 sensor is used. Ten language options.
Delivery information: Supplied with PT100 sensor, standard spindles (four for the L model and six for the R and H models), viscometer stand and spindle protector, carrying case, USB cable and data logger software.