L(+)-Serine

Synonyms: Serine(S)-(+)-SerineH-Ser-OH(S)-2-Amino-3-hydroxypropanoic acid

EDQMS0450000EA 231 EUR
EDQMS0450000
L(+)-Serine
L(+)-Serine
Formula: C₃H₇NO₃
MW: 105,09 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 215…225 °C
Density: 1,6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR