Iron(III) chloride hexahydrate 97.0-102.0% (by iodometry), lumps, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor
Danger

Synonyms: Iron trichloride hexahydrateFerric chloride hexahydrate

C5029-20EA 5458.78 EUR
C5029-20
Iron(III) chloride hexahydrate 97.0-102.0% (by iodometry), lumps, AR® ACS, Macron Fine Chemicals™
Iron (III) chloride hexahydrate
Formula: FeCl₃·6H₂O
MW: 270,3 g/mol
Boiling Pt: 280…285 °C (1013 hPa)
Melting Pt: 37 °C
Density: 1,82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://ie.vwr.com/stibo/web/std.lang.all/05/51/16640551.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (FeCl₃·6H₂O) (by iodometry) 97.0 - 102.0 %
Solubility Passes Test
Identification A Passes Test
Identification B Passes Test
ACS - Calcium (Ca)(by FES) ≤0.01 %
ACS - Copper (Cu) ≤0.003 %
ACS - Insoluble Matter ≤0.01 %
Ferrous Iron ≤0.02 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Nitrate (NO₃) ≤0.01 %
ACS - Phosphorus Compounds (as PO₄) ≤0.01 %
ACS - Potassium (K) ≤0.005 %
ACS - Sodium (Na) ≤0.05 %
ACS - Sulfate (SO₄) ≤0.01 %
ACS - Zinc (Zn) ≤0.003 %
Appearance (brown-yellow crystalline lumps)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR