Boron nitride ≥99.5% (metals basis), rods low binder, Length: 203mm, Ø 6.4mm
Supplier: Thermo Scientific
45573.KGEA
234
EUR
45573.KG
Boron nitride ≥99.5% (metals basis), rods low binder, Length: 203mm, Ø 6.4mm
Boron nitride
Formula:
BN MW: 24,82 g/mol Melting Pt: 2527 °C Density: 2,15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011317 CAS Number: 10043-11-5 EINECS: 233-136-6 Merck Index: 13,01336 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...