Clindamycin hydrochloride, >835 µg/mg, white solid

Supplier: BioVision

Synonyms: (2S,4R)-N-((1S,2S)-2-Chloro-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochlorideCleocin Hydrochloride7[S]-Chloro-7-deoxylincomycin

BIOV2627-50EA 333 EUR
BIOV2627-50 BIOV2627-10
Clindamycin hydrochloride, >835 µg/mg, white solid
Clindamycin hydrochloride

Clindamycin is a broad spectrum antibiotic targeting primarily gram positive and gram negative bacteria such as Clostridium and Bacteroides species


It inhibits bacterial protein synthesis by binding the 50S ribosomal subunit and interfering with tRNA activity during translation.


Solubility: H₂O(~ 50 mg/ml)


Caution: For Research Use Only. Not for use in humans.
Protect from air and moisture.

Formula: C₁₈H₃₃ClN₂O₅S·HCl
MW: 461,45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD07793327
CAS Number: 21462-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR