You Searched For: Cellulose Stoppers
Protecting microbiology samples and tissue cultures from bacteria, the cellulose stoppers snugly fit in test tubes or Erlenmeyer flasks. Routinely used in laboratories, the breathable closures permit with air flow for when a complete tight seal is not necessary. Due to the organic compound manufacturing and hydrophilic physical properties, closures are for single use only. The standard, thick, or specialized shaped cellulose stoppers come in different diameters to fit any opening needed.
Protecting microbiology samples and tissue cultures from bacteria, the cellulose stoppers snugly fit in test tubes or Erlenmeyer flasks. Routinely used in laboratories, the breathable closures permit with air flow for when a complete tight seal is not necessary. Due to the organic compound manufacturing and hydrophilic physical properties, closures are for single use only. The standard, thick, or specialized shaped cellulose stoppers come in different diameters to fit any opening needed.
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR Collection
Description: These stoppers are made of cellulose and are used for microbiological samples and tissue cultures in test tubes or Erlenmeyer flasks.
Supplier: VWR Collection
Description: These plugs are made of bonded dense weave cellulose acetate which provides an excellent barrier to mite infestation. They are precision manufactured ...
Supplier: HERENZ, HEINZ GMBH
Description: Original Steristoppers® have been proved as an excellent sealing for microbiological samples and tissue cultures in test tubes and Erlenmeyer flasks. ...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 3 of 3
no targeter for Bottom