Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Compressed Air Stations

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"3"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (PEAK42-1030UK)

Supplier:  PEAK SCIENTIFIC INSTRUMENTS LT
Description:   TOC gas generators produce up to 1500 ml/min of carrier/combustion gas from in house compressed air supply. They are recommended for NPOC (Non-purgeab...
UOM:  1 * 1 items

Supplier:  PEAK SCIENTIFIC INSTRUMENTS LT
Description:   The Precision air compressor is designed specifically to provide a dedicated, fully integrated compressed air source for Precision Nitrogen and Precis...
UOM:  1 * 1 items
Catalog Number: (PEAK65-0003)

Supplier:  PEAK SCIENTIFIC INSTRUMENTS LT
Description:   This air compressor generator has been designed to support the Solaris nitrogen generator for ELSD in laboratories without an external supply of high ...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 3  of 3
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping