Disinfectant spray, Soft Care Des E
Supplier: Diversey
DIVY7515520EA 0 EUR
DIVY7515520
Disinfectant spray, Soft Care Des E
Cleaning Agents Disinfectants
Ethanol-based hand disinfection product.
  • Does not require water and can therefore be used everywhere
  • Ethanol is very resistant against bacteria, moulds, yeasts and viruses
Soft Care Des E also contains a moisturising agent for skin care.
Certifications: Meets the European standards for disinfection (including EN 1500 standard).
Caution: Use biocides safely. Always read the label and product information before use.