4-[(5-Aminopyridin-2-yl)oxy]benzoic acid

Supplier: Apollo Scientific

OR103637-1G OR103637-250MG
APOSOR103637-1GEA 306 EUR
APOSOR103637-1G APOSOR103637-250MG
4-[(5-Aminopyridin-2-yl)oxy]benzoic acid
4-[(5-Aminopyridin-2-yl)oxy]benzoic acid
Formula: C₁₂H₁₀N₂O₃
MW: 230,219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445671
CAS Number: 887350-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR