4-(tert-Butyldimethylsilyloxy)methylbenzhydrazide

Supplier: Apollo Scientific
Warning

OR110422-1G OR110422-10G OR110422-5G
APOSOR110422-1GEA 193 EUR
APOSOR110422-1G APOSOR110422-10G APOSOR110422-5G
4-(tert-Butyldimethylsilyloxy)methylbenzhydrazide
4-(tert-Butyldimethylsilyloxy)methylbenzhydrazide
Formula: C₁₄H₂₄N₂O₂Si
MW: 280,438 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18804149
CAS Number: 179055-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR