2-(4-Methylhomopiperazin-1-yl)benzoic acid hydrochloride hemihydrate

Supplier: Apollo Scientific
Warning

OR12216-1G OR12216-5G
APOSOR12216-1GEA 263 EUR
APOSOR12216-1G APOSOR12216-5G
2-(4-Methylhomopiperazin-1-yl)benzoic acid hydrochloride hemihydrate
2-(4-Methylhomopiperazin-1-yl)benzoic acid hydrochloride hemihydrate
Formula: C₁₃H₁₈N₂O₂·ClH·H₂O
MW: 288,77 g/mol
Melting Pt: 167…173 °C
Storage Temperature: Ambient
MDL Number: MFCD09258630
CAS Number: 921938-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR