3-[(Pyridin-2-yl)oxy]benzaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: 2-(3-Formylphenoxy)pyridine

OR12893-1G OR12893-250MG
APOSOR12893-1GEA 270 EUR
APOSOR12893-1G APOSOR12893-250MG
3-[(Pyridin-2-yl)oxy]benzaldehyde
3-[(Pyridin-2-yl)oxy]benzaldehyde
Formula: C₁₂H₉NO₂
MW: 199,205 g/mol
Melting Pt: 115…116 °C
Storage Temperature: Ambient
MDL Number: MFCD03840110
CAS Number: 137386-78-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR