4-Isocyanato-4-(thien-2-yl)tetrahydro-2H-pyran 95%

Supplier: Apollo Scientific
Danger

Synonyms: 2-(4-Isocyanatotetrahydro-2H-pyran-4-yl)thiophene,4-(Thien-2-yl)tetrahydro-2H-pyran-4-yl isocyanate

OR14870-1G OR14870-250MG
APOSOR14870-1GEA 212 EUR
APOSOR14870-1G APOSOR14870-250MG
4-Isocyanato-4-(thien-2-yl)tetrahydro-2H-pyran 95%
4-Isocyanato-4-(thien-2-yl)tetrahydro-2H-pyran

When melted, may contain small amount of sediment

Formula: C₁₀H₁₁NO₂S
MW: 209,265 g/mol
Melting Pt: 49,5…52.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09879931
CAS Number: 926921-63-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR