5-Amino-1,3-dihydrobenzo[c]furan

Supplier: Apollo Scientific
Warning

Synonyms: 1,3-Dihydroisobenzofuran-5-amine,5-Amino-1,3-dihydro-2-benzofuran,1,3-Dihydro-2-benzofuran-5-amine

OR16559-1G OR16559-250MG
APOSOR16559-1GEA 384 EUR
APOSOR16559-1G APOSOR16559-250MG
5-Amino-1,3-dihydrobenzo[c]furan
5-Amino-1,3-dihydrobenzo[c]furan
Formula: C₈H₉NO
MW: 135,163 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD05864561
CAS Number: 61964-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR