1,2-di(chloromethyl)-4,5-dimethylbenzene

Supplier: Apollo Scientific

OR21094-10G OR21094-25G
APOSOR21094-10GEA 154 EUR
APOSOR21094-10G APOSOR21094-25G
1,2-di(chloromethyl)-4,5-dimethylbenzene
1,2-di(chloromethyl)-4,5-dimethylbenzene
Formula: C₁₀H₁₂Cl₂
MW: 203,108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000911
CAS Number: 2362-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR