5-Bromo-3,4-dimethylbenzene-1,2-diamine

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-3,4-dimethylphenylene-1,2-diamine,6-Bromo-3,4-diamino-o-xylene

OR21478-1G OR21478-250MG OR21478-5G
APOSOR21478-1GEA 94.5 EUR
APOSOR21478-1G APOSOR21478-250MG APOSOR21478-5G
5-Bromo-3,4-dimethylbenzene-1,2-diamine
5-Bromo-3,4-dimethylbenzene-1,2-diamine
Formula: C₈H₁₁BrN₂
MW: 215,09 g/mol
Melting Pt: 87…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00052696
CAS Number: 107100-16-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR