5-(4-Methoxyphenyl)-1-methylpyridin-2(1H)-one

Supplier: Apollo Scientific

OR2281-100MG OR2281-500MG
APOSOR2281-100MGEA 102 EUR
APOSOR2281-100MG APOSOR2281-500MG
5-(4-Methoxyphenyl)-1-methylpyridin-2(1H)-one
5-(4-Methoxyphenyl)-1-methylpyridin-2(1H)-one
Formula: C₁₃H₁₃NO₂
MW: 215,248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907673
CAS Number: 945980-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR