4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride
Supplier: Apollo Scientific
APOSOR350394-1GEA
94.5
EUR
APOSOR350394-1G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride
Formula:
C₁₁H₂₀BNO₂·ClH MW: 245,554 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11506076 CAS Number: 1121057-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...