7-Bromo-2-phenylquinoline 95+%

APOSOR350432-5GEA 0 EUR
APOSOR350432-5G APOSOR350432-250MG APOSOR350432-1G
7-Bromo-2-phenylquinoline 95+%
7-Bromo-2-phenylquinoline
Formula: C₁₅H₁₀BrN
MW: 284,151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702481
CAS Number: 1203578-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR