1,4-Benzodioxane-5-boronic acid 95+%

Supplier: Apollo Scientific

Synonyms: (2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)boronic acid

APOSOR360091-1GEA 601 EUR
APOSOR360091-1G APOSOR360091-250MG APOSOR360091-100MG
1,4-Benzodioxane-5-boronic acid 95+%
1,4-Benzodioxane-5-boronic acid
Formula: C₈H₉BO₄
MW: 179,966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095151
CAS Number: 499769-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR