3-(Methoxymethoxy)phenylboronic acid 95%

Supplier: Apollo Scientific

APOSOR360231-1GEA 147 EUR
APOSOR360231-1G APOSOR360231-5G
3-(Methoxymethoxy)phenylboronic acid 95%
3-(Methoxymethoxy)phenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181,982 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040407
CAS Number: 216443-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR