2-Morpholinopyrimidin-5-ylboronic acid 95%

Supplier: Apollo Scientific

Synonyms: 2-(Morpholine-4-yl)pyrimidine-5-boronic acid

APOSOR360280-250MGEA 75.7 EUR
APOSOR360280-250MG
2-Morpholinopyrimidin-5-ylboronic acid 95%
2-Morpholinopyrimidin-5-ylboronic acid
Formula: C₈H₁₂BN₃O₃
MW: 209,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375148
CAS Number: 870521-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR