1-Isobutyl-1H-pyrazole-4-boronic acid 98%
Supplier: Apollo Scientific
Synonyms:
(1-Isobutyl-1H-pyrazol-4-yl)boronic acid
APOSOR360330-1GEA
166
EUR
APOSOR360330-1G
1-Isobutyl-1H-pyrazole-4-boronic acid 98%
1-Isobutyl-1H-pyrazole-4-boronic acid
Formula:
C₇H₁₃BN₂O₂ MW: 168,001 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09951916 CAS Number: 929094-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...