4-(2-Hydroxyethyl)phenylboronic acid 97%
Supplier: Apollo Scientific
Synonyms:
(4-(2-Hydroxyethyl)phenyl)boronic acid
APOSOR361251-250MGEA
115
EUR
APOSOR361251-250MG
4-(2-Hydroxyethyl)phenylboronic acid 97%
4-(2-Hydroxyethyl)phenylboronic acid
Formula:
C₈H₁₁BO₃ MW: 165,982 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095135 CAS Number: 137756-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...