3-Chloro-1-methyl-1H-pyrazol-4-amine ≥98%

Supplier: Apollo Scientific

APOSOR370032-100MGEA 368 EUR
APOSOR370032-100MG
3-Chloro-1-methyl-1H-pyrazol-4-amine ≥98%
3-Chloro-1-methyl-1H-pyrazol-4-amine
Formula: C₄H₆ClN₃
MW: 131,564 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19214410
CAS Number: 150187-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR