2-Benzyl-2,5-diazaspiro[3.4]octan-1-one ≥98%

Supplier: Apollo Scientific

APOSOR370049-100MGEA 970 EUR
APOSOR370049-100MG
2-Benzyl-2,5-diazaspiro[3.4]octan-1-one ≥98%
2-Benzyl-2,5-diazaspiro[3.4]octan-1-one
Formula: C₁₃H₁₆N₂O
MW: 216,279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18207189
CAS Number: 1415562-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR