2-Oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepine-8-carbonitrile ≥98%
Supplier: Apollo Scientific
APOSOR370060-100MGEA
1210
EUR
APOSOR370060-100MG
2-Oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepine-8-carbonitrile ≥98%
2-Oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepine-8-carbonitrile
Formula:
C₁₁H₁₀N₂O MW: 186,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18642821 CAS Number: 903557-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...