3-Iodo-2-methylquinolin-4-ol ≥98%

Supplier: Apollo Scientific

APOSOR370083-1GEA 1640 EUR
APOSOR370083-1G
3-Iodo-2-methylquinolin-4-ol ≥98%
3-Iodo-2-methylquinolin-4-ol
Formula: C₁₀H₈INO
MW: 285,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368411
CAS Number: 64965-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR