1,3-Dichloro-5,6-dihydro-4H-cyclopenta[c]thiophen-4-one 97%

APOSOR370103-250MGEA 0 EUR
APOSOR370103-250MG
1,3-Dichloro-5,6-dihydro-4H-cyclopenta[c]thiophen-4-one 97%
1,3-Dichloro-5,6-dihydro-4H-cyclopenta[c]thiophen-4-one
Formula: C₇H₄Cl₂OS
MW: 207,077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094480
CAS Number: 7687-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR