(4-(2-Chlorophenyl)thiazol-2-yl)methanol 95%

APOSOR370112-1GEA 0 EUR
APOSOR370112-1G
(4-(2-Chlorophenyl)thiazol-2-yl)methanol 95%
(4-(2-Chlorophenyl)thiazol-2-yl)methanol
Formula: C₁₀H₈ClNOS
MW: 225,695 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17251190
CAS Number: 1050507-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR