6-Hydroxy-1H-indole-3-carbaldehyde 98%

Supplier: Apollo Scientific

APOSOR370135-250MGEA 1210 EUR
APOSOR370135-250MG
6-Hydroxy-1H-indole-3-carbaldehyde 98%
6-Hydroxy-1H-indole-3-carbaldehyde
Formula: C₉H₇NO₂
MW: 161,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110391
CAS Number: 192184-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR