3-(4-Ethoxybenzoyl)acrylic acid 98%

Supplier: Apollo Scientific
Warning

APOSOR4000-1GEA 14.7 EUR
APOSOR4000-1G
3-(4-Ethoxybenzoyl)acrylic acid 98%
3-(4-Ethoxybenzoyl)acrylic acid
Formula: C₁₂H₁₂O₄
MW: 220,221 g/mol
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00075337
CAS Number: 29582-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR