Methyl 2-chloro-1,3-benzoxazole-6-carboxylate 95%
Supplier: Apollo Scientific
|
Warning
|
APOSOR400449-250MGEA
568
EUR
APOSOR400449-250MG
Methyl 2-chloro-1,3-benzoxazole-6-carboxylate 95%
Methyl 2-chloro-1,3-benzoxazole-6-carboxylate
Formula:
C₉H₆ClNO₃ MW: 211,602 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD20528144 CAS Number: 819076-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...